missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-6 Human Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of human IL-6 protein levels from samples including serum, plasma, and cell culture supernatants.

£172.00 - £566.00

Specifications

Accession Number P05231
Assay Range 2-200 pg/mL
Assay Sensitivity 2 pg/mL
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Alert:

Contains Azide


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15551037
View Documents
Invitrogen™
88-7066-86
10 x 96 Tests
£408.00
Pack of 10
In Stock
to see stock.
Add to basket
 
15531037
View Documents
Invitrogen™
88-7066-76
20 x 96 Tests
£566.00
Pack of 20
In Stock
to see stock.
Add to basket
 
15521037
View Documents
Invitrogen™
88-7066-22
2 x 96 Tests
£172.00
Pack of 2
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

The Human Interleukin-6 (IL-6) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-6. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Human IL-6 ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ IL-6 Human Uncoated ELISA Kit with Plates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.