missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-6 QuantiGlo ELISA Kit
Human IL-6 QuantiGlo ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ Q6000B
This item is not returnable.
View return policy
Description
The Human IL-6 QuantiGlo ELISA Kit quantifies human IL-6 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).Specifications
0.48 to 1500 pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human IL-6 | |
Human | |
3569 | |
1 Plate | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
IL-6 |
0.35 pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Cell Culture Supernatant, Citrate, Plasma (EDTA, Citrate, Heparin), Plasma (EDTA, Citrate, Heparin), Serum, Serum | |
Sufficient materials to run ELISAs on at least one 96-well plate | |
ELISA | |
IL6 | |
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL) | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction