missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-7 Recombinant Protein
Recombinant Human Interleukin 7 (IL7) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC0075
This item is not returnable.
View return policy
Description
Human IL-7 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human PBMCs
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Approved for shipment on Wet or Dry Ice.
Specifications
Cell Culture | |
17.5 kDa | |
5μg | |
Human | |
Carrier-Free | |
5 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
3574 | |
Sequential Chromatography | |
IL-7 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Signal Transduction, Stem Cell Research, Drug Development, Oncology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.