missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-8 ELISA Kits

Product Code. p-4531774
Click to view available options
Quantity:
1 Plate Kit
5 Plate Kit
Sufficient For:
5 x 96 microplate wells
96 microplate wells
Sample Type:
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Urine
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10167393

Brand: Invitrogen™ EH2IL8

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Interleukin 8 Recombinant protein SIL8 from Thermo Scientific™ specifically detects Interleukin 8 in Not Applicable samples, and it is validated for Control, ELISA and WB

Assay IL-8 (human interleukin-8) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples with our Invitrgen™ Human IL-8 ELISA Kits.

These human IL-8 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-8 (IL-8) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IL-8 antibody pairs and IL-8 standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human interleukin-8 (IL-8) and has been shown to recognize IL-8 derived from stimulated human umbilical vein endothelial cells

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 25.6 to 1000pg/mL

  • Sensitivity: < 2pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with with human PF-4, GROα, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-6, IL-7, IL-10, IL-12, TNFα, IFNα, IFNγ, or GM-CSF

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 25.6 to 1000pg/mL
Assay Sensitivity <2pg/mL
Description Human IL-8 ELISA Kit
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity No significant cross-reaction with with human PF-4, GROa, IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-6, IL-7, IL-10, IL-12, TNFa, IFNa, IFNg, or GM-CSF
Sufficient For 96 microplate wells
Kit Contents Streptavidin-HRP Concentrate, 75 μL, TMB Substrate Solution, 13mL, Stop Solution, 13mL, Standard Diluent, 25mL, 30X Wash Buffer, 50mL, Lyophilized Recombinant Human IL-8 Standard, 2 vials, Adhesive Plate Covers, 6 ea, Anti-Human IL-8 Precoated 96-well Strip Plate, 1 ea, Biotinylated Antibody Reagent, 8mL, HRP Dilution Buffer, 14mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 1 Plate Kit
Sample Volume 50 μL
Target Natural and recombinant human interleukin-8 (IL-8) and has been shown to recognize IL-8 derived from stimulated human umbilical vein endothelial cells
Total Assay Time 3 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.