missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human INHA Recombinant Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
This product contains purified recombinant human Inhibin a protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone.
Specifications
Specifications
| Accession Number | P05111 |
| Concentration | 0.16 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | tris HCl with 50% glycerol, 5 mM EDTA and no preservative; pH 8 |
| Gene ID (Entrez) | 3623 |
| Name | Human INHA |
| pH Range | 8 |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction