missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRF8 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
IRF8 Synthetic Peptide, PEP-0206, from Invitrogen.
IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.
Specifications
Specifications
| Gene ID (Entrez) | 3394 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q02556 |
| Common Name | Human IRF8 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction