Learn More
Invitrogen™ Human ITIH1 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP301
Additional Details : Weight : 0.99000kg
Product Code. 10540101
  • £154.00 / Each
Estimated Shipment: 13-08-2024
to see stock.
Description
Description
ITI-HC1 Synthetic Peptide, PEP-301, from Invitrogen™.
The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]). This peptide corresponds to amino acids Q(383) VQESLPELSN (393) of human ITI-HC1 and can be used as a blocking peptide with polyclonal antibody PA1-1755.
Specifications
Specifications
3697
purified
Human ITIH1
-20°C
Lyophilized
Blocking Assay, Control
ITIH1
No preservative
50 μg
Human
P19827
1 mg/mL
Synthetic Peptide
water with no preservative
CYCSP39, cytochrome c, somatic pseudogene 39, HCP39
Yes
Peptide
RUO - research use only