missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Keratinocyte Growth Supplement (HKGS)
Human Keratinocyte Growth Supplement (HKGS)
Brand: Gibco™ S0015
46.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
A sterile, concentrated (100X) solution intended for use with EpiLife™ medium or Medium 154 to culture human epidermal keratinocytes. HKGS is an ionically-balanced supplement containing bovine pituitary extract (BPE), recombinant human insulin-like growth factor-I, hydrocortisone, bovine transferrin, and human epidermal growth factor. Each 5 mL bottle of HKGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human epidermal keratinocytes, and is the correct amount of supplement for a 500 mL bottle of medium.When a 500 mL bottle of medium is supplemented with HKGS, the final concentrations of the components in the supplemented medium are:
- Bovine pituitary extract (BPE), 0.2% v/v
- Recombinant human insulin-like growth factor-I, 0.01 μg/mL
- Hydrocortisone, 0.18μg/mL
- Bovine transferrin, 5μg/mL
- Human epidermal growth factor, 0.2 ng/mL

Specifications
Store in freezer (-5 to -20°C). | |
100 X | |
Growth Supplement | |
Mammalian Cell Culture, Primary Cell Culture | |
5 mL | |
Human |
Keratinocyte Cells | |
Liquid | |
Sterile | |
Cascade Biologics™ | |
Human |
User Guide: Human Keratinocyte Growth Supplement (HKGS)
Version: 3.0
File size: 114.4 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Human Keratinocyte Growth Supplement (HKGS) > 5mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction