Learn More
Invitrogen™ Human Kininogen ELISA Kit
Human Kininogen quantitates human Kininogen in serum, plasma, supernatant.
Brand: Invitrogen™ EH287RBX10
Additional Details : Weight : 2.50000kg
Description
Human Kininogen quantitates human Kininogen in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Kininogen.
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.Specifications
Alpha-1-MAP; alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; high molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; Kng; KNG1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; Williams-Fitzgerald-Flaujeac factor | |
24 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
KNG1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
24 to 6,000 ng/mL | |
Biotin | |
Serum | |
P01042 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 µL; Serum, 50 µL; Plasma, 50 µL | |
Kininogen 1 | |
4 hr 45 min |