Learn More
Invitrogen™ Human Lass4 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP12926
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Lass4 Synthetic Peptide, PEP-12926, from Invitrogen.
It is thought that Lass4 may be either a bona fide (dihydro)ceramide synthase or a modulator of its activity. When overexpressed in cells is involved in the production of sphingolipids containing different fatty acid donnors (N-linked stearoyl- (C18) or arachidoyl- (C20) ceramides) in a fumonisin B1-independent manner.Specifications
79603 | |
purified | |
Human Lass4 | |
Synthetic Peptide | |
PBS with 0.08% sodium azide | |
CERS4 | |
0.08% sodium azide | |
50 μg | |
PBS |
Human | |
Q9HA82 | |
-20°C. Avoid freeze-thaw cycles. | |
Liquid | |
Control, Western Blot | |
No | |
Peptide | |
RUO - research use only |
in vitro research only.