missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LDLR Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0866

Product Code. 15399100

  • £112.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

LDL-R Synthetic Peptide, PEP-0866, from Invitrogen.

This peptide corresponds to 18 amino acids near the center of human LDL-R.PEP-0866 can be used as a blocking peptide with polyclonal antibody PA5-20752.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3949
purified
Human LDLR
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
P01130
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
LDLR
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers