Learn More
R&D Systems™ Human LGI1 (NP_005088) VersaClone cDNA
R&D Systems™ Human LGI1 (NP_005088) VersaClone cDNA is a member of the secreted leucine-rich repeat (LRR) superfamily.
Brand: R&D Systems™ RDC2017
This item is not returnable.
View return policy
Description
The Human LGI1 (NP_005088) VersaClone cDNA clone contains the complete ORF for Lgi1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Lgi1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
hLGI1 | |
10 μg at 0.2 μg/μL | |
1687bp | |
9211 | |
Human |
NP_005088 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |