missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human LIF Recombinant Protein

Recombinant human leukemia inhibitory factor (LIF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9481

Code : NEW

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 13330595

  • £2510.00 / 100µg
Estimated Shipment: 16-09-2024
to see stock.

Description

Description

Human LIF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

LIF is a cytokine that induces macrophage differentiation. LIF, in conjunction with BMP2, acts on primary fetal neural progenitor cells to induce astrocytes.
  • High purity: minimizes interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the LIF dose-dependent proliferation of Human TF-1 cells

Shipping Condition: Dry Ice.

Specifications

Specifications

21 kDa
100μg
LIF
Untagged
0.5 ng/ml
Dry Ice
Cytokines & Receptors
Lyophilized
Oncology
95 %
Sequential Chromatography
LIF
Human
Lyophilized form should be stored at 2–8°C, preferably desiccated.
Reconstituted form should be stored at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
Dose-dependent proliferation of TF-1 cells
E. coli
Recombinant, Ligand
Other Cytokines
Gibco™
Leukemia inhibitory factor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.