missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human LIGHT Recombinant Protein, PeproTech®
Insect cells Recombinant Protein
£64.75 - £3190.00
Specifications
Accession Number | O43557 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 8740 |
Molecular Weight (g/mol) | 19.3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17892403
|
Gibco™
310-09B-100UG |
7 x 15 μg | N/A | ||||||
17882233
|
Gibco™
310-09B-15UG |
15 μg |
£157.00
15µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17822413
|
Gibco™
310-09B-1MG |
67 x 15 μg | N/A | ||||||
17822033
|
Gibco™
310-09B-250UG |
17 x 15 μg | N/A | ||||||
17881993
|
Gibco™
310-09B-3UG |
3 μg |
£64.75
3µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17852003
|
Gibco™
310-09B-500UG |
34 x 15 μg | N/A | ||||||
Description
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.Specifications
O43557 | |
protein with no preservative | |
19.3 | |
Insect cells | |
RUO | |
CD258; Herpes virus entry mediator ligand; herpesvirus entry mediator ligand; HVEML; HVEM-L; Light; LTg; Ly113; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tnfsf14; TR2; tumor necrosis factor (ligand) superfamily, member 14; tumor necrosis factor ligand 1 D; tumor necrosis factor ligand superfamily member 14; Tumor necrosis factor ligand superfamily member 14, membrane form; Tumor necrosis factor ligand superfamily member 14, soluble form; tumor necrosis factor superfamily member 14; UNQ391/PRO726 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
8740 | |
Human LIGHT | |
-20°C | |
<1 EU/μg | |
TNFSF14 | |
Unconjugated | |
RLGEMVTRLP DGPAGSWEQL IQERRSHEVN PAAHLTGANS SLTGSGGPLL WETQLGLAFL RGLSYHDGAL VVTKAGYYYI YSKVQLGGVG CPLGLASTIT HGLYKRTPRY PEELELLVSQ QSPCGRATSS SRVWWDSSFL GGVVHLEAGE KVVVRVLDER LVRLRDGTRS YFGAFMV | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |