missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB1/CD85j/ILT2 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
£147.94 - £383.36
Specifications
Antigen | ILT2/CD85j/LILRB1 |
---|---|
Clone | 292305 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730577
|
R&D Systems
MAB20171-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18193281
|
R&D Systems
MAB20171 |
100 μg |
£383.36
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILT2/CD85j/LILRB1 Monoclonal specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
ILT2/CD85j/LILRB1 | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q8NHL6 | |
10859 | |
Mouse myeloma cell line NS0-derived recombinant human LILRB1/CD85j/ILT2 Gly24-His458 Accession # Q8NHL6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human LILRB1/CD85j/ILT2 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ILT4, rhILT5, rhLIR6 or rhLIR8 is observed. |
292305 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD85 antigen-like family member J, CD85J, CD85j antigen, Ig-like transcript 2, ILT2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1, LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, MIR7, Monocyte/macrophage immunoglobulin-like receptor 7 | |
LILRA5 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only