missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human M-CSF, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£177.00 - £3755.00
Specifications
Accession Number | P09603 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 1435 |
Molecular Weight (g/mol) | 36.8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17833063
|
Gibco™
AF-300-25-100UG |
100 μg |
£778.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17883553
|
Gibco™
AF-300-25-10UG |
10 μg |
£177.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17813803
|
Gibco™
AF-300-25-1MG |
1 mg |
£3755.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17813383
|
Gibco™
AF-300-25-500UG |
500 μg |
£2255.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.Specifications
P09603 | |
protein with no preservative | |
36.8 | |
E. coli | |
RUO | |
C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
1435 | |
Human M-CSF, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
Csf1 | |
Unconjugated | |
MEEVSEYCSH MIGSGHLQSL QRLIDSQMET SCQITFEFVD QEQLKDPVCY LKKAFLLVQD IMEDTMRFRD NTPNAIAIVQ LQELSLRLKS CFTKDYEEHD KACVRTFYET PLQLLEKVKN VFNETKNLLD KDWNIFSKNC NNSFAECSSQ GHERQSEGS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |