missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAGP-2/MFAP5 ELISA Kit
Human MAGP-2/MFAP5 quantitates human MAGP-2/MFAP5 in serum, plasma, supernatant.
£540.00 - £3870.00
Specifications
Accession Number | AAT9; MAGP2; MAGP-2; MFAP5; MFAP-5; Microfibril-associated glycoprotein 2; microfibril-associated glycoprotein-2; microfibrillar associated protein 5; microfibrillar-associated protein 5; MP25; THE1A-MFAP5 |
---|---|
Assay Range | 0.04 to 10 ng/mL |
Assay Sensitivity | 0.04 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17195079
|
Invitrogen™
EH312RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17844120
|
Invitrogen™
EH312RBX10 |
10 x 96 Tests |
£3870.00
Pack of 10 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
17104613
|
Invitrogen™
EH312RB |
96 Tests |
£540.00
Pack of 96 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
Description
Human MAGP-2/MFAP5 quantitates human MAGP-2/MFAP5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MAGP-2/MFAP5.
This gene encodes a 25-kDa microfibril-associated glycoprotein which is rich in serine and threonine residues. It lacks a hydrophobic carboxyl terminus and proline-, glutamine-, and tyrosine-rich regions, which are characteristics of a related 31- kDa microfibril-associated glycoprotein (MFAP2). The close similarity between these two proteins is confined to a central region of 60 aa where precise alignment of 7 cysteine residues occurs. The structural differences suggest that this encoded protein has some functions that are distinct from those of MFAP2.Specifications
AAT9; MAGP2; MAGP-2; MFAP5; MFAP-5; Microfibril-associated glycoprotein 2; microfibril-associated glycoprotein-2; microfibrillar associated protein 5; microfibrillar-associated protein 5; MP25; THE1A-MFAP5 | |
0.04 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
MFAP5 | |
<10% | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
MFAP5 | |
4 hr 45 min |
0.04 to 10 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q13361 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |