missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Melanocyte Growth Supplement (HMGS)

Product Code. 10378213
Click to view available options
Quantity:
5 mL
missing translation for 'unitSize'
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10378213

missing translation for 'mfr': Gibco™ S0025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Human Melanocyte Growth Supplement (HMGS)

A sterile, concentrated (100X) solution intended for use with Medium 254 or Medium 254CF to culture human epidermal melanocytes. Each 5 ml bottle of HMGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human epidermal melanocytes and is the correct amount of supplement for a 500 ml bottle of basal Medium 254 or Medium 254CF.

When a 500 ml bottle of medium is supplemented with HMGS, the final concentrations of the components in the supplemented medium are:

  • Bovine pituitary extract (BPE) (0.2% v/v)
  • Fetal bovine serum (0.5% v/v)
  • Recombinant human insulin-like growth factor-I (1μg/ml)
  • Bovine transferrin (5μg/ml)
  • Basic fibroblast growth factor (3 ng/ml)
  • Hydrocortisone (0.18μg/ml)
  • Heparin (3μg/ml)
  • Phorbol 12-myristate 13-acetate (PMA) (10 ng/ml)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer (-5 to -20°C).
    Cell Type Mammalian Cells
    Concentration 100 X
    Form Liquid
    Product Type Growth Supplement
    Sterility Sterile
    Product Line Cascade Biologics
    Quantity 5 mL
    Shipping Condition Dry Ice
    Source Human
    Species Human
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.