Learn More
Invitrogen™ Human Meprin alpha ELISA Kit
Human Meprin alpha quantitates human Meprin alpha in serum, plasma, supernatant.
Brand:  Invitrogen™ EH321RB
 View more versions of this product
Product Code. 17104623
  • £540.00 / Pack of 96
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Human Meprin alpha quantitates human Meprin alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Meprin alpha.
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; Endopeptidase-24.18 subunit alpha; Mep1; MEP-1; Mep1a; Mep-1 A; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; Meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA
2 ng/mL
ELISA
Colorimetric Microplate Reader
MEP1A
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
2 to 400 ng/mL
Biotin
Plasma, Serum, Supernatant
Q16819
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL
MEP1A
4 hr 45 min