missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Meprin alpha ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EH321RBX10

Product Code. 17707703

  • £3950.00 / Pack of 10
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Q16819
2 ng/mL
ELISA
Colorimetric Microplate Reader
MEP1A
<10%
HRP
RUO
2°C to 8°C
1 hr 20 min
2 to 400 ng/mL
Biotin
Plasma, Serum, Supernatant
4224
<12%
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers
10 x 96 Tests
Supernatant, 50 μL|Serum, 50 μL|Plasma, 50 μL
MEP1A
4 hr 45 min
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers