Learn More
STEMCELL Technologies Human Mesenchymal Stem Cell qPCR Array
For characterization of human MSCs and their differentiation to chondrogenic, adipogenic, or osteogenic lineages
Brand:  STEMCELL Technologies 07543
Encompass_Preferred
 View more versions of this product
Product Code. 17189291
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • The Human Mesenchymal Stem Cell Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of human mesenchymal stem and progenitor cells (MSCs) and their differentiated chondrogenic, adipogenic, or osteogenic progeny
  • MSCs can be expanded in vitro to generate mesenchymal stromal cell cultures, which, under appropriate conditions, can differentiate into adipocytes, chondrocytes, and osteoblasts
  • This qPCR array provides the gene expression profile of MSCs and their trilineage derivatives following in vitro differentiation
Specifications
Specifications
Adipocytes; Chondrocytes; Mesenchymal Stem and Progenitor Cells; Osteoblasts
96-well Plate Type 3
Real Time PCR (qPCR)