Learn More
STEMCELL Technologies Human Mesenchymal Stem Cell qPCR Array
For characterization of human MSCs and their differentiation to chondrogenic, adipogenic, or osteogenic lineages
Brand: STEMCELL Technologies 07544
This item is not returnable.
View return policy
Description
- The Human Mesenchymal Stem Cell Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of human mesenchymal stem and progenitor cells (MSCs) and their differentiated chondrogenic, adipogenic, or osteogenic progeny
- MSCs can be expanded in vitro to generate mesenchymal stromal cell cultures, which, under appropriate conditions, can differentiate into adipocytes, chondrocytes, and osteoblasts
- This qPCR array provides the gene expression profile of MSCs and their trilineage derivatives following in vitro differentiation
Specifications
Adipocytes; Chondrocytes; Mesenchymal Stem and Progenitor Cells; Osteoblasts | |
96-well Plate Type 4 | |
Real Time PCR (qPCR) |