Learn More
R&D Systems™ Human mGluR1 (AAB05337) VersaClone cDNA
Glutamate receptor, metabotropic 1
Brand:  R&D Systems™ RDC0983
Product Code. 18701229
  • £1025.00 / 10µg
Explore more special offers
Description
Description
The Human mGluR1 (AAB05337) VersaClone cDNA clone contains the complete ORF for mGluR1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The mGluR1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans