missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human MIP-3 (CCL23) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 300-29-100UG
This item is not returnable.
View return policy
Description
This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.Specifications
P55773 | |
protein with no preservative | |
11.3 | |
100 μg | |
-20°C | |
<1 EU/μg | |
CCL23 | |
Unconjugated | |
RVTKDAETEF MMSKLPLENP VLLDRFHATS ADCCISYTPR SIPCSLLESY FETNSECSKP GVIFLTKKGR RFCANPSDKQ VQVCMRMLKL DTRIKTRKN | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
6368 | |
Human MIP-3 (CCL23) | |
E. coli | |
RUO | |
C6 beta-chemokine; C-C motif chemokine 23; C-C motif chemokine ligand 23; CCL23; CCL23(19-99); CCL23(22-99); CCL23(27-99); CCL23(30-99); chemokine (C-C motif) ligand 23; Ck beta 8-1; CKb8; CKB-8; Ckb-8-1; CK-BETA-8; hmrp-2 A; Macrophage inflammatory protein 3; MIP3; MIP-3; MIP-3 CK-BETA-8; MPIF1; MPIF-1; MPIF-1 Ckb-8-1; Myeloid progenitor inhibitory factor 1; SCYA23; small inducible cytokine subfamily A (Cys-Cys), member 23; Small-inducible cytokine A23 | |
Human | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction