missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human MMP-1 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 420-01-2UG
60.22 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.Specifications
P03956 | |
protein with no preservative | |
42.7 | |
2 μg | |
-20°C | |
<1 EU/μg | |
MMP1 | |
Unconjugated | |
MFVLTEGNPR WEQTHLTYRI ENYTPDLPRA DVDHAIEKAF QLWSNVTPLT FTKVSEGQAD IMISFVRGDH RDNSPFDGPG GNLAHAFQPG PGIGGDAHFD EDERWTNNFR EYNLHRVAAH ELGHSLGLSH STDIGALMYP SYTFSGDVQL AQDDIDGIQA IYGRSQNPVQ PIGPQTPKAC DSKLTFDAIT TIRGEVMFFK DRFYMRTNPF YPEVELNFIS VFWPQLPNGL EAAYEFADRD EVRFFKGNKY WAVQGQNVLH GYPKDIYSSF GFPRTVKHID AALSEENTGK TYFFVANKYW RYDEYKRSMD PGYPKMIAHD FPGIGHKVDA VFMKDGFFYF FHGTRQYKFD PKTKRILTLQ KANSWFNCRK N | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
4312 | |
Human MMP-1 | |
E. coli | |
RUO | |
22 kDa interstitial collagenase; 27 kDa interstitial collagenase; CLG; CLGN; Collagenase; collagenase-like A; fibroblast collagenase; Interstitial collagenase; interstitial collagenase A; matrix metallo protease; matrix metallopeptidase 1; matrix metallopeptidase 1 (interstitial collagenase); matrix metallopeptidase 1 A; matrix metallopeptidase 1 A (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; matrix metalloproteinase 1 A (interstitial collagenase); Matrix metalloproteinase1; Matrix metalloproteinase-1; Matrix metalloproteinase-1 A; McolA; Mcol-A; MMP; MMP1; MMP-1; Mmp1a; MMP-1 A; MMPs | |
Human | |
Recombinant | |
Lyophilized |