missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse CD45R / B220 Rat, Biotin, Clone: RA3-6B2, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB41-B-500ug

 View more versions of this product

Product Code. 15941988

  • £155.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Human/Mouse CD45R / B220
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2a κ
Flow Cytometry
RA3-6B2
Biotin
NP_001255215
Rat
500 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers