missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD45R / B220 Rat, PE-Cy7, Clone: RA3-6B2, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB41-PE7-25ug
This item is not returnable.
View return policy
Specifications
Human/Mouse CD45R / B220 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ptprc | |
Protein A/G Purified | |
RUO | |
19264 | |
IgG2a κ |
Flow Cytometry | |
RA3-6B2 | |
PE-Cyanine7 | |
NP_001255215 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |