Learn More
Gibco™ Human/Mouse FGF-8b Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 100-25-250UG
 View more versions of this product
Product Code. 17800293
  • £838.00 / 250µg
Explore more special offers
Description
Description
FGF-8 (FGF-8b) is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8A, FGF-8B, FGF-8E and FGF-8F. The human and murine FGF-8A and B are identical, unlike human and mouse FGF-8E and F, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P37237, P55075
protein with no preservative
22.5
250 μg
-20°C
<1 EU/μg
Human, Mouse
Recombinant
Lyophilized
Functional Assay
14179, 2253
Human/Mouse FGF-8 b
E. coli
RUO
FGF8
Unconjugated
MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR
≥ 95% by SDS-PAGE gel and HPLC analyses.