missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human/Mouse/Rat BDNF, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£177.00 - £4550.00
Specifications
Accession Number | P21237, P23363, P23560 |
---|---|
For Use With (Application) | ELISA, Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 12064, 24225, 627 |
Molecular Weight (g/mol) | 27 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17803283
|
Gibco™
AF-450-02-100UG |
100 μg |
£946.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17853363
|
Gibco™
AF-450-02-10UG |
10 μg |
£177.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17883443
|
Gibco™
AF-450-02-1MG |
1 mg |
£4550.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17813143
|
Gibco™
AF-450-02-500UG |
500 μg |
£2730.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.Specifications
P21237, P23363, P23560 | |
protein with no preservative | |
27 | |
E. coli | |
RUO | |
abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF | |
Human, Mouse, Rat | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay | |
12064, 24225, 627 | |
Human/Mouse/Rat BDNF, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
BDNF | |
Unconjugated | |
MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |