missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human/Mouse/Rat Irisin Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
Brand: Gibco™ 100-65-50UG
This item is not returnable.
View return policy
Description
Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.Specifications
Q8K3V5, Q8K4Z2, Q8NAU1 | |
protein with no preservative | |
25.2 | |
50 μg | |
-20°C | |
<1 EU/μg | |
FNDC5 | |
Unconjugated | |
DSPSAPVNVT VRHLKANSAV VSWDVLEDEV VIGFAISQQK KDVRMLRFIQ EVNTTTRSCA LWDLEEDTEY IVHVQAISIQ GQSPASEPVL FKTPREAEKM ASKNKDEVTM KE | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
252995, 260327, 384061 | |
Human/Mouse/Rat Irisin | |
CHO cells | |
RUO | |
1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp | |
Human, Mouse, Rat | |
Recombinant | |
Lyophilized |