Learn More
Invitrogen™ Human/Mouse/Rat SERCA3 ATPase (aa 29-39) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP015
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 1.24740kg
Description
Mouse, Rat, Human SERCA3 (residues 29-39) Synthetic Peptide, PEP-015, from Invitrogen.
PEP-015 is an 11 amino acid (amino acids 29-39) synthetic peptide whose sequence is from the amino end of SERCA3 that is conserved in mouse, rat, and human SERCA3. The sequence of this peptide is (amino to carboxy terminus): V-T-D-A-R-E-R-Y-G-P-NThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and serca3, catalog # PA1-910A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Specifications
25391, 489, 53313 | |
purified | |
Human/Mouse/Rat SERCA3 ATPase (aa 29-39) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human, Mouse, Rat | |
P18596, Q64518, Q93084 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
ATP2A3 | |
No preservative | |
50 μg | |
V - T - D - A - R - E - R - Y - G - P - N |