missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TREM2 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 13 publications
Brand: R&D Systems MAB17291-SP
This item is not returnable.
View return policy
Description
TREM2 Monoclonal specifically detects TREM2 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry, CyTOF-ready, Knockout Validated.Specifications
TREM2 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, ELISA, Immunohistochemistry 0.5-15 ug/mL, Immunocytochemistry 5-25 ug/mL, CyTOF-ready, Knockout Validated | |
Q99NH8 | |
TREM2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREM-2 extracellular domain Accession # Q99NH8 | |
25 μg | |
Primary | |
Detects human and mouse TREM-2 in direct ELISAs and Western blots. Stains TREM-2 transfectants but not TREM-1 transfectants. | |
Human, Mouse | |
Purified |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, CyTOF, Immunoassay | |
237920 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
54209 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only