Learn More
R&D Systems™ Human Myeloid Dendritic Cell Multi-Color Flow Kit
Designed for the flow cytometric analysis of mDCs using four fluorochrome-conjugated antibodies
Brand:  R&D Systems™ FMC016
Additional Details : Weight : 0.09700kg
Product Code. 18752084
  • £835.83 / 1 set
Estimated Shipment: 17-07-2024
to see stock.
Description
Description
Single-step staining
Conjugated antibodies (and corresponding isotype controls): BDCA1/CD1c-APC (goat IgG), BDCA3/CD141-PE (Clone 501733; mouse IgG1), CD16-PerCP (Clone 245536; mouse IgG2A), CD11c-CFS (Clone ICRF 3.9; mouse IgG1)
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.