Learn More
Invitrogen™ Human MZT1 (aa 61-82) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP103998
154.00 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64105 (PA5-64105. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Required for gamma-tubulin complex recruitment to the centrosome.
Specifications
Q08AG7 | |
Blocking Assay, Control | |
440145 | |
100 μL | |
2410129H14Rik; AI120158; C13orf37; mitotic spindle organizing protein 1; mitotic-spindle organizing protein 1; Mitotic-spindle organizing protein associated with a ring of gamma-tubulin 1; MOZART1; MZT1; si:ch211-141i4.2; si:dkey-15d12.2 | |
MZT1 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human MZT1 (aa 61-82) Control Fragment | |
RUO | |
MZT1 | |
Unconjugated | |
Recombinant | |
SSVIKELRKATEALKAAENMTS | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.