missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Nectin-2/CD112 ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EH331RBX10

Product Code. 17767703

  • £3950.00 / Pack of 10
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Q92692
0.05 ng/mL
ELISA
Colorimetric Microplate Reader
NECTIN2
<10%
HRP
RUO
2°C to 8°C
1 hr 20 min
0.05 to 10 ng/mL
Biotin
Plasma, Serum, Supernatant
5819
<12%
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers
10 x 96 Tests
Supernatant, 50 μL|Serum, 50 μL|Plasma, 50 μL
Nectin 2
4 hr 45 min
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers