missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Netrin-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB1254
This item is not returnable.
View return policy
Description
Netrin-4 Monoclonal specifically detects Netrin-4 in Human samples. It is validated for Western Blot.Specifications
Netrin-4 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9HB63 | |
NTN4 | |
Mouse myeloma cell line NS0-derived recombinant human Netrin-4 Val26-Lys628 Accession # Q9HB63 | |
500 μg | |
Primary | |
Detects human Netrin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Netrin-1, recombinant chicken (rch) Netrin-1, rchNetrin-2, or rmNetrin-G1a is observed. | |
Human | |
Purified |
Western Blot | |
191633 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta-netrin, hepar-derived netrin-like protein, netrin 4, Netrin4, NTN4, PRO3091 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
59277 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
For Research Use Only