missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Neurturin Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 450-11-250UG
This item is not returnable.
View return policy
Description
Glial cell line-derived neurotrophic factor (GDNF) plays key roles in the control of vertebrate neuron survival and differentiation. A novel neurotrophic factor was recently cloned from human and mouse and designated neurturin. Physiological responses to neurturin (NTN) require the presence of receptor tyrosine kinase RET and a novel glycosylphosphatidylinositol linked receptor NTNRalpha. The cDNAs encoding NTNRalpha from human, rat, chicken, and mouse have been cloned recently and termed GDNFR-b, Ret ligand 2 (RETL2) or TGF-beta-related neurotrophic factor receptor 2 (TrnR2) and nominated as GFR-a-2 recently. NTN binds to and forms a complex with GFR-a-2 and the Ret PTK and activates the RET receptor tyrosine kinase pathway. Both NTN and GDNF can activate the MAP kinase and phosphatidylinositol 3-kinase pathways and play a critical role in the development of many neuronal populations. Neurturin and GDNF define a new family of neurotrophic factors.Specifications
Q99748 | |
protein with no preservative | |
23.6 | |
250 μg | |
-20°C | |
<1 EU/μg | |
NRTN | |
Unconjugated | |
ARLGARPCGL RELEVRVSEL GLGYASDETV LFRYCAGACE AAARVYDLGL RRLRQRRRLR RERVRAQPCC RPTAYEDEVS FLDAHSRYHT VHELSARECA CV | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
4902 | |
Human Neurturin | |
E. coli | |
RUO | |
neurturin; NRTN; NTN; prepro-neurturin | |
Human | |
Recombinant | |
Lyophilized |