missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neutrophil Elastase/ELA2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£189.00 - £479.00
Specifications
Antigen | Neutrophil Elastase/ELA2 |
---|---|
Clone | 950317R |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 0.05-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18712087
|
R&D Systems
MAB91671R-SP |
25 μg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18771517
|
R&D Systems
MAB91671R-100 |
100 μg |
£479.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
Neutrophil Elastase/ELA2 | |
Western Blot 1 ug/mL, Immunohistochemistry 0.05-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
Chinese hamster ovary cell line CHO-derived recombinant human Neutrophil Elastase/ELA2, Met1-Asn252, Accession # P08246 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Neutrophil Elastase/ELA2 in direct ELISAs. |
950317R | |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P08246 | |
1991 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |