Learn More
Invitrogen™ Human NRF2 ELISA Kit
Human NRF2 quantitates human NRF2 in serum, plasma, supernatant.
Brand: Invitrogen™ EH348RBX10
Additional Details : Weight : 2.50000kg
Description
Human NRF2 quantitates human NRF2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NRF2.
Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.Specifications
AI194320; E4TF1-60; E4TF1A; HEBP1; NFE2L2; NF-E2-related factor 2; NFE2-related factor 2; NFT2; NRF2; NRF2A; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2 | |
14 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
NFE2L2 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
14 to 10,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q16236 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
Nrf2 | |
4 hr 45 min |