Learn More
Gibco™ Human OATP1B1 (G) SLC Transporter Cells
HEK293 cells that transiently overexpress the OATP1B1 solute carrier (SLC) transport protein
Brand: Gibco™ GM1102G
636.50 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- TRANSiPORT Human OATP1B1 (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the OATP1B1 solute carrier (SLC) transport protein. Solute carriers are a super-family of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes in and out of cells. The OATP1B1 SLC transporter protein is found in hepatocytes.
- Assess potential for transporter-mediated drug metabolism
- Easy to use format
- Obtain high quality results with a large signal-to-noise ratio
- Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
- Mock HEK293 cells (Cat. No. GM1101G) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of transport due to the presence of endogenous transporters or non-specific binding.
- The convenient product format enables data generation in 2 days from thawing of the cells to final results.
Specifications
Store in liquid nitrogen vapor phase. | |
Mammalian Cell Culture | |
8-10 x 106 cells | |
8–10M cells |
293 (HEK) | |
Human | |
SLC Transporter Cell |