missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Oncostatin M (209 aa) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 300-10T-2UG
43.16 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Oncostatin M (OSM) is a growth and differentiation factor that participates in the regulation of neurogenesis, osteogenesis and hematopoiesis. Produced by activated T cells, monocytes and Kaposi's sarcoma cells, OSM can exert both stimulatory and inhibitory effects on cell proliferation. It stimulates the proliferation of fibroblasts, smooth muscle cells and Kaposi's sarcoma cells, but inhibits the growth of some normal and tumor cell lines. It also promotes cytokine release (e.g. IL-6, GM-CSF and G-CSF) from endothelial cells, and enhances the expression of low-density lipoprotein receptors in hepatoma cells. OSM shares several structural and functional characteristics with LIF, IL-6, and CNTF.Specifications
P13725 | |
protein with no preservative | |
23.6 | |
2 μg | |
-20°C | |
<1 EU/μg | |
OSM | |
Unconjugated | |
AAIGSCSKEY RVLLGQLQKQ TDLMQDTSRL LDPYIRIQGL DVPKLREHCR ERPGAFPSEE TLRGLGRRGF LQTLNATLGC VLHRLADLEQ RLPKAQDLER SGLNIEDLEK LQMARPNILG LRNNIYCMAQ LLDNSDTAEP TKAGRGASQP PTPTPASDAF QRKLEGCRFL HGYHRFMHSV GRVFSKWGES PNRSRRHSPH QALRKGVRR | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
5008 | |
Human Oncostatin M (209 aa) | |
E. coli | |
RUO | |
MGC20461; OncoM; oncostatin M; oncostatin-M; OSM | |
Human | |
Recombinant | |
Lyophilized |