missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Oncostatin M (227 aa) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £4150.00
Specifications
Accession Number | P13725 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 5008 |
Molecular Weight (g/mol) | 25.7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17881603
|
Gibco™
300-10-100UG |
100 μg |
£832.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17891003
|
Gibco™
300-10-10UG |
10 μg |
£157.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17820243
|
Gibco™
300-10-1MG |
1 mg |
£4150.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17871203
|
Gibco™
300-10-250UG |
250 μg |
£1456.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17800633
|
Gibco™
300-10-2UG |
2 μg |
£64.75
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861713
|
Gibco™
300-10-500UG |
500 μg |
£2495.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831603
|
Gibco™
300-10-50UG |
50 μg |
£502.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Oncostatin M (OSM) is a growth and differentiation factor that participates in the regulation of neurogenesis, osteogenesis and hematopoiesis. Produced by activated T cells, monocytes and Kaposi's sarcoma cells, OSM can exert both stimulatory and inhibitory effects on cell proliferation. It stimulates the proliferation of fibroblasts, smooth muscle cells and Kaposi's sarcoma cells, but inhibits the growth of some normal and tumor cell lines. It also promotes cytokine release (e.g. IL-6, GM-CSF and G-CSF) from endothelial cells, and enhances the expression of low-density lipoprotein receptors in hepatoma cells. OSM shares several structural and functional characteristics with LIF, IL-6, and CNTF.Specifications
P13725 | |
protein with no preservative | |
25.7 | |
E. coli | |
RUO | |
MGC20461; OncoM; oncostatin M; oncostatin-M; OSM | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
5008 | |
Human Oncostatin M (227 aa) | |
-20°C | |
<1 EU/μg | |
OSM | |
Unconjugated | |
AAIGSCSKEY RVLLGQLQKQ TDLMQDTSRL LDPYIRIQGL DVPKLREHCR ERPGAFPSEE TLRGLGRRGF LQTLNATLGC VLHRLADLEQ RLPKAQDLER SGLNIEDLEK LQMARPNILG LRNNIYCMAQ LLDNSDTAEP TKAGRGASQP PTPTPASDAF QRKLEGCRFL HGYHRFMHSV GRVFSKWGES PNRSRRHSPH QALRKGVRRT RPSRKGKRLM TRGQLPR | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |