missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human OSCAR (NP_573399) VersaClone cDNA
Osteoclast-associated Receptor
Brand: R&D Systems™ RDC1948
Additional Details : Weight : 0.00970kg
Description
The Human OSCAR (NP_573399) VersaClone cDNA clone contains the complete ORF for OSCAR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The OSCAR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
cDNA | |
NP_573399 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Human |
hOSCAR | |
10 μg at 0.2 μg/μL | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
For Research Use Only