Learn More
Novus Biologicals™ Human Ovary Membrane Tissue Lysate (Adult Membrane Normal)
Brand: Novus Biologicals™ NB820-60603
Additional Details : Weight : 0.00970kg
Description
The Human Ovary Membrane Tissue Lysate (Adult Membrane Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Specifications
Western Blot | |
0.1 mg | |
Membrane Protein - Human Adult Normal Tissue: Ovary | |
Normal | |
Human | |
Human Ovary Membrane Tissue Lysate (Adult Normal) | |
Adult |
Reproductive | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Membrane | |
Human | |
1.3 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
For Research Use Only