missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human P-Cadherin ELISA Kit

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous P-cadherin

Brand:  Proteintech™ KE00117-96T


Product Code. 16851526

  • £219.00 / 96 tests
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Recognizes human P-cadherin
  • Cadherins are a family of transmembrane glycoproteins that mediate calcium-dependent cell-cell adhesion and play role in maintenance of normal tissue architecture
    • P-cadherin (Placental cadherin, CDH3, cadherin-3) is a classical member of cadherin superfamily
    • Expression reported in many cancers (breast, gastric, endometrial, ovian and prostate)
    • Can be cleaved into soluble fragment, found in human  milk, semen, serum and NAF

Process:

  • Antibody specific for P-cadherin pre-coated onto microwells
  • P-cadherin protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody of biotinylated specific for P-cadherin is added to detect captured P-cadherin protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications

Specifications

BC041846
0.5 pg/mL
Sandwich ELISA, Double Antibody
Plasma, Serum
Human
Human
<10%
Blue ice
4.5 hr.
31.25 to 2000 pg/mL
ISO 13485
Coated ELISA Kit
DAB
1001
Solid
<10%
-20°C to +4°C
P22223
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 96TEST Human P-cadherin ELISA Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.