Learn More
Invitrogen™ Human Parkin (aa 298-313) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP159
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human Parkin (residues 298-313) Synthetic Peptide, PEP-159, from Invitrogen.
PEP-159 is a 15 amino acid synthetic peptide whose sequence is from human parkin. The sequence of this peptide is (amino to carboxy terminus): C- I (298)-K-E-L-H-H-F-R-I-L- G-E-E-Q-Y-N (313)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human parkin, catalog # PA1-751. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Specifications
5071 | |
purified | |
Human Parkin (aa 298-313) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
O60260 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
PRKN | |
No preservative | |
50 μg | |
C- I(298)-K-E-L-H-H-F-R-I-L- G-E-E-Q-Y-N(313) |