Learn More
Gibco™ Human PDGF-AA Recombinant Protein
Human PDGF-AA Recombinant Protein
Brand: Gibco™ PHG0035
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human PDGF-AA Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Members of the PDGF family are mitogenic factors for cells of mesenchymal origin.- High purity: nno interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the PDGF dose dependent proliferation of BALB/c 3T3 fibroblasts
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Especificaciones
Cell Culture | |
28.5 kDa | |
10 ug | |
250 aa (homodimer) | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Stem Cell Research, Angiogenesis, Oncology | |
98 % |
5154 | |
Sequential Chromatography | |
PDGF-AA | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
PDGFs (Platelet-Derived Growth Factors) | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.