missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human PDGF-BB Recombinant Protein

Recombinant Human Platelet Derived Growth Factor-BB (PDGF-BB) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHG0044

 View more versions of this product

Product Code. 10531285

  • £116.00 / 5µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Human PDGF-BB Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

Members of the PDGF family are mitogenic factors for cells of mesenchymal origin.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/c 3T3 cells

Shipping Condition: Wet Ice.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Culture
25.4 kDa
5μg
Human
Carrier-Free
5 μg
E. coli
Full Length, Recombinant, Ligand
PDGFs (Platelet-Derived Growth Factors)
Gibco™
5155
Sequential Chromatography
PDGF-BB
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Approved for shipment on Wet or Dry Ice
Growth Factors & Receptors
Lyophilized
Immunology, Neurobiology, Stem Cell Research, Angiogenesis, Oncology
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.