missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human PDGF-BB Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £3110.00
Specifications
Accession Number | P01127 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 5155 |
Molecular Weight (g/mol) | 24.3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17880383
|
Gibco™
100-14B-100UG |
100 μg |
£622.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17850403
|
Gibco™
100-14B-10UG |
10 μg |
£157.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17839912
|
Gibco™
100-14B-1MG |
1 mg |
£3110.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870883
|
Gibco™
100-14B-250UG |
250 μg |
£1090.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831333
|
Gibco™
100-14B-2UG |
2 μg |
£64.75
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880083
|
Gibco™
100-14B-500UG |
500 μg |
£1868.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17821023
|
Gibco™
100-14B-50UG |
50 μg |
£375.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
P01127 | |
protein with no preservative | |
24.3 | |
E. coli | |
RUO | |
becaplermin; c-sis; FLJ12858; H-PDGF-BB; IBGC5; PDGF; pdgf protein; PDGF subunit B; PDGF, B chain; PDGF2; PDGF-2; PDGFB; PDGF-B; platelet derived growth factor subunit B; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor B; platelet-derived growth factor B chain; platelet-derived growth factor beta; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); proto-oncogene c-Sis; Sis; SSV | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
5155 | |
Human PDGF-BB | |
-20°C | |
<1 EU/μg | |
PDGFB | |
Unconjugated | |
SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPIFKKATVT LEDHLACKCE TVAAARPVT | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title