missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Pepsin ELISA Kit
For the quantitative detection of Human Pepsin
Brand: Invitrogen™ EEL057
Additional Details : Weight : 0.40000kg
Description
PGA5: Pepsinogen 5, group I (pepsinogen A). Pepsinogens are the inactive precursors of pepsin, the major acid protease found in the stomach. Pepsin is one of the main proteolytic enzymes secreted by the gastric mucosa. Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity; although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. PGA5 is a member of the subfamily A1 within the pepsin family and is the predominant endopeptidase in the gastric juice of vertebrates. PGA5 is inhibited by ovUS-1, a uterine serpin.Specifications
P0DJD9 | |
37.5 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Pga5 | |
<10% | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hr 20 min |
62.5 to 4000 pg/mL | |
HRP | |
Serum | |
5222 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 Tests | |
100 µL | |
Human | |
3 hr 30 min |